Skip to Content
Merck
4′-Bromoacetophenone 98%

4′-Bromoacetophenone

4′-Bromoacetophenone 98%
Synonym(s):
1-Acetyl-4-bromobenzene, 4′-Bromoacetophenone
Linear Formula:
4-(Br)C6H4COCH3
CAS Number:
Molecular Weight:
199.04

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing