![4-Amino-4H-1,2,4-triazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/210/116/8b416622-e24e-4f6b-a229-e8163bfbe015/640/8b416622-e24e-4f6b-a229-e8163bfbe015.png)
4-Amino-4H-1,2,4-triazole
![4-Amino-4H-1,2,4-triazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/210/116/8b416622-e24e-4f6b-a229-e8163bfbe015/640/8b416622-e24e-4f6b-a229-e8163bfbe015.png)
Synonym(s):
4H-1,2,4-Triazol-4-amine
Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2