![4-Amino-2,6-dichloropyrimidine 95%](/deepweb/assets/sigmaaldrich/product/structures/313/513/ab818195-3225-424d-9603-1fc51f772e71/640/ab818195-3225-424d-9603-1fc51f772e71.png)
4-Amino-2,6-dichloropyrimidine
![4-Amino-2,6-dichloropyrimidine 95%](/deepweb/assets/sigmaaldrich/product/structures/313/513/ab818195-3225-424d-9603-1fc51f772e71/640/ab818195-3225-424d-9603-1fc51f772e71.png)
Empirical Formula (Hill Notation):
C4H3Cl2N3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing