![(3R,6R)-3,6-Diphenylpiperazine-2,5-dione certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/images/725/339/dda30848-846f-438d-a098-3b5a0419b7f9/640/dda30848-846f-438d-a098-3b5a0419b7f9.jpg)
(3R,6R)-3,6-Diphenylpiperazine-2,5-dione
![(3R,6R)-3,6-Diphenylpiperazine-2,5-dione certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/images/725/339/dda30848-846f-438d-a098-3b5a0419b7f9/640/dda30848-846f-438d-a098-3b5a0419b7f9.jpg)
Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing