Skip to Content
Merck
3-Pyrrolidinol 98%

3-Pyrrolidinol

3-Pyrrolidinol 98%
Synonym(s):
3-Hydroxypyrrolidine
Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing