Skip to Content
Merck
3-Chloro-4-morpholin-4-yl-1,2,5-thiadiazole 98%

3-Chloro-4-morpholin-4-yl-1,2,5-thiadiazole

3-Chloro-4-morpholin-4-yl-1,2,5-thiadiazole 98%
Synonym(s):
4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine
Empirical Formula (Hill Notation):
C6H8ClN3OS
CAS Number:
Molecular Weight:
205.67
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing