Skip to Content
Merck
3,5-Dimethoxybenzoyl chloride 97%

3,5-Dimethoxybenzoyl chloride

3,5-Dimethoxybenzoyl chloride 97%
Linear Formula:
(CH3O)2C6H3COCl
CAS Number:
Molecular Weight:
200.62
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing