Skip to Content
Merck
3,4-Dichlorophenol 99%

3,4-Dichlorophenol

3,4-Dichlorophenol 99%
Synonym(s):
3,4-Dichlorophenol
Linear Formula:
3,4-(Cl)2C6H3OH
CAS Number:
Molecular Weight:
163.00

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing