Skip to Content
Merck
(−)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 99%

(−)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate

(−)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 99%
Synonym(s):
α-Methyl-L-DOPA, L-α-Methyl-DOPA
Linear Formula:
(HO)2C6H3CH2C(CH3)(NH2)CO2H · 11/2H2O
CAS Number:
Molecular Weight:
238.24
EC Number:

Products

Shipping
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1