![2-Thiazoline-2-thiol 98%](/deepweb/assets/sigmaaldrich/product/structures/234/856/c4409878-dbc6-4400-9078-efe4f6294293/640/c4409878-dbc6-4400-9078-efe4f6294293.png)
2-Thiazoline-2-thiol
![2-Thiazoline-2-thiol 98%](/deepweb/assets/sigmaaldrich/product/structures/234/856/c4409878-dbc6-4400-9078-efe4f6294293/640/c4409878-dbc6-4400-9078-efe4f6294293.png)
Synonym(s):
2-Mercapto-2-thiazoline, 2-Thiazoline-2-thiol
Empirical Formula (Hill Notation):
C3H5NS2
CAS Number:
Molecular Weight:
119.21
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing