2-(methylthio)pyrimidin-4-amine
Empirical Formula (Hill Notation):
C5H7N3S
CAS Number:
Molecular Weight:
141.19
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing