Skip to Content
Merck
2-(Methylthio)-2-thiazoline 97%

2-(Methylthio)-2-thiazoline

2-(Methylthio)-2-thiazoline 97%
Synonym(s):
2-(Methylmercapto)-2-thiazoline
Empirical Formula (Hill Notation):
C4H7NS2
CAS Number:
Molecular Weight:
133.24
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing