Skip to Content
Merck
2-Methyl-4-oxopentane-2-thiol ≥99%, FG

2-Methyl-4-oxopentane-2-thiol

2-Methyl-4-oxopentane-2-thiol ≥99%, FG
Synonym(s):
4-Methyl-4-mercaptopentan-2-one
Empirical Formula (Hill Notation):
C6H12OS
CAS Number:
Molecular Weight:
132.22

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing