![2-Methyl-3-furanthiol natural, 1% in Sunflower oil](/deepweb/assets/sigmaaldrich/product/structures/193/132/98c510d6-c11f-4ebb-9719-cb20fe02ceba/640/98c510d6-c11f-4ebb-9719-cb20fe02ceba.png)
2-Methyl-3-furanthiol
![2-Methyl-3-furanthiol natural, 1% in Sunflower oil](/deepweb/assets/sigmaaldrich/product/structures/193/132/98c510d6-c11f-4ebb-9719-cb20fe02ceba/640/98c510d6-c11f-4ebb-9719-cb20fe02ceba.png)
Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
Products
Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing