2-Mesitylmagnesium bromide solution
Synonym(s):
2,4,6-Trimethylphenylmagnesium bromide
Linear Formula:
(CH3)3C6H2MgBr
CAS Number:
Molecular Weight:
223.39
Products
Shipping
Brand
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing