![2-Mesitylenesulfonic acid dihydrate 97%](/deepweb/assets/sigmaaldrich/product/structures/371/840/3f25b723-9f0c-4efd-9812-900c87a6fb54/640/3f25b723-9f0c-4efd-9812-900c87a6fb54.png)
2-Mesitylenesulfonic acid dihydrate
![2-Mesitylenesulfonic acid dihydrate 97%](/deepweb/assets/sigmaaldrich/product/structures/371/840/3f25b723-9f0c-4efd-9812-900c87a6fb54/640/3f25b723-9f0c-4efd-9812-900c87a6fb54.png)
Synonym(s):
2,4,6-Trimethylbenzenesulfonic acid dihydrate
Linear Formula:
(CH3)3C6H2SO3H · 2H2O
CAS Number:
Molecular Weight:
236.29
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing