Skip to Content
Merck
2′-Fucosyllactose ≥95% (NMR)

2′-Fucosyllactose

2′-Fucosyllactose ≥95% (NMR)
Synonym(s):
2′-Fucosyllactose, 2′-Ο-Fucosyllactose
Empirical Formula (Hill Notation):
C18H32O15
CAS Number:
Molecular Weight:
488.44

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing