Skip to Content
Merck
2-Cyano-6-methoxybenzothiazole 99%

2-Cyano-6-methoxybenzothiazole

2-Cyano-6-methoxybenzothiazole 99%
Synonym(s):
6-Methoxy-2-benzothiazolecarbonitrile
Empirical Formula (Hill Notation):
C9H6N2OS
CAS Number:
Molecular Weight:
190.22

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing