
2-Chloro-N,N-dimethylpropylamine hydrochloride

Synonym(s):
β-(Dimethylamino)isopropyl chloride hydrochloride, 2-(Dimethylamino)isopropyl chloride hydrochloride, 2-Chloro-1-(dimethylamino)propane hydrochloride, N-(2-Chloropropyl)dimethylamine hydrochloride
Linear Formula:
CH3CHClCH2N(CH3)2 · HCl
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing