Skip to Content
Merck
2-Chloro-6-iodobenzoic acid 95%

2-Chloro-6-iodobenzoic acid

2-Chloro-6-iodobenzoic acid 95%
Empirical Formula (Hill Notation):
C7H4ClIO2
CAS Number:
Molecular Weight:
282.46

Products

Shipping
Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing