
2-Chloro-4-(trifluoromethyl)aniline

Synonym(s):
4-Amino-3-chlorobenzotrifluoride
Linear Formula:
ClC6H3(CF3)NH2
Products
Shipping
Product Category
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing