Skip to Content
Merck
2-Benzylphenol ≥98.0% (GC)

2-Benzylphenol

2-Benzylphenol ≥98.0% (GC)
Synonym(s):
α-Phenyl-o-cresol, 2-Hydroxydiphenylmethane
Linear Formula:
C6H5CH2C6H4OH
CAS Number:
Molecular Weight:
184.23
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing