![2-Amino-6-hydroxybenzothiazole 95%](/deepweb/assets/sigmaaldrich/product/structures/385/141/8c350559-d09d-48e1-af35-903dceda1b1d/640/8c350559-d09d-48e1-af35-903dceda1b1d.png)
2-Amino-6-hydroxybenzothiazole
![2-Amino-6-hydroxybenzothiazole 95%](/deepweb/assets/sigmaaldrich/product/structures/385/141/8c350559-d09d-48e1-af35-903dceda1b1d/640/8c350559-d09d-48e1-af35-903dceda1b1d.png)
Synonym(s):
2-Amino-6-benzothiazolol, 6-Hydroxy-2-aminobenzothiazole, 6-Hydroxybenzothiazol-2-amine
Empirical Formula (Hill Notation):
C7H6N2OS
CAS Number:
Molecular Weight:
166.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing