Skip to Content
Merck
2-Amino-4,5-imidazoledicarbonitrile 97%

2-Amino-4,5-imidazoledicarbonitrile

2-Amino-4,5-imidazoledicarbonitrile 97%
Empirical Formula (Hill Notation):
C5H3N5
CAS Number:
Molecular Weight:
133.11
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing