
2-Amino-2-thiazoline

Synonym(s):
2-Amino-2-thiazoline
Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing