Skip to Content
Merck
2,5-Hexanedione ≥98%

2,5-Hexanedione

2,5-Hexanedione ≥98%
Synonym(s):
Acetonylacetone, 2,5-Hexanedione
Linear Formula:
CH3COCH2CH2COCH3
CAS Number:
Molecular Weight:
114.14

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing