![2,4-Dichlorobenzyl alcohol impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/413/145/30f5fbbb-91dc-496a-a25d-72ea72a55780/640/30f5fbbb-91dc-496a-a25d-72ea72a55780.png)
2,4-Dichlorobenzyl alcohol impurity A
![2,4-Dichlorobenzyl alcohol impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/413/145/30f5fbbb-91dc-496a-a25d-72ea72a55780/640/30f5fbbb-91dc-496a-a25d-72ea72a55780.png)
Synonym(s):
2,5-Dichlorobenzyl alcohol
Linear Formula:
Cl2C6H3CH2OH
CAS Number:
Molecular Weight:
177.03
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing