Skip to Content
Merck
2-(2-Hydroxy-5-methylphenyl)benzotriazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-(2-Hydroxy-5-methylphenyl)benzotriazole

2-(2-Hydroxy-5-methylphenyl)benzotriazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(2H-Benzotriazol-2-yl)-p-cresol
Empirical Formula (Hill Notation):
C13H11N3O
CAS Number:
Molecular Weight:
225.25

Products

Shipping
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reference Material Type
Special Grade