
2,2-Dipropylpentanoic acid

Synonym(s):
2,2-Dipropylvaleric acid, Tripropylacetic acid
Empirical Formula (Hill Notation):
C11H22O2
CAS Number:
Molecular Weight:
186.29
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing