2,2-Dimethoxyacetaldehyde solution
Synonym(s):
Glyoxal 1-(dimethyl acetal), Glyoxal dimethyl acetal
Linear Formula:
(CH3O)2CHCHO
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing