![{2-[(2,6-Dichlorophenyl)amino]phenyl}methanol pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/122/595/f97308b7-b691-4671-a20d-5c784ad41402/640/f97308b7-b691-4671-a20d-5c784ad41402.png)
{2-[(2,6-Dichlorophenyl)amino]phenyl}methanol
![{2-[(2,6-Dichlorophenyl)amino]phenyl}methanol pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/122/595/f97308b7-b691-4671-a20d-5c784ad41402/640/f97308b7-b691-4671-a20d-5c784ad41402.png)
Synonym(s):
2-[(2,6-Dichlorophenyl)amino]benzenemethanol, o-(2,6-Dichloroanilino)benzyl alcohol
Empirical Formula (Hill Notation):
C13H11Cl2NO
CAS Number:
Molecular Weight:
268.14
Products
Shipping
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing