Skip to Content
Merck
2-[(2,6-Dichlorophenyl)amino]benzaldehyde pharmaceutical impurity standard

2-[(2,6-Dichlorophenyl)amino]benzaldehyde

2-[(2,6-Dichlorophenyl)amino]benzaldehyde pharmaceutical impurity standard
Synonym(s):
2-(2,6-Dichloroanilino)benzaldehyde, N-(2,6-Dichlorophenyl)anthranilaldehyde
Empirical Formula (Hill Notation):
C13H9Cl2NO
CAS Number:
Molecular Weight:
266.12

Products

Shipping
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing