![2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥96.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/237/185/b94eadd2-5a2c-4a5b-a13d-3d3905df0dbc/640/b94eadd2-5a2c-4a5b-a13d-3d3905df0dbc.png)
2-[2-(2-Aminoethoxy)ethoxy]ethanol
![2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥96.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/237/185/b94eadd2-5a2c-4a5b-a13d-3d3905df0dbc/640/b94eadd2-5a2c-4a5b-a13d-3d3905df0dbc.png)
Synonym(s):
2-[2-(2-Hydroxyethoxy)ethoxy]ethylamine, 3,6,9-Trioxadecylamine, 8-Amino-3,6-dioxaoctanol, NSC 40760, T3EGMA, Triethylene glycol monoamine, Triglycolamine
Empirical Formula (Hill Notation):
C6H15NO3
CAS Number:
Molecular Weight:
149.19
Products
Shipping
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1