1-Propanol-d8
Synonym(s):
Deuterated Propanol, Deuterated propyl alcohol, Propyl alcohol-d8
Linear Formula:
CD3CD2CD2OD
CAS Number:
Molecular Weight:
68.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing