Skip to Content
Merck
1-Methylnaphthalene analytical standard

1-Methylnaphthalene

1-Methylnaphthalene analytical standard
Synonym(s):
1-Methylnaphthalene
Linear Formula:
C10H7CH3
CAS Number:
Molecular Weight:
142.20
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing