Skip to Content
Merck
1-Methyl-3-phenylpropylamine 98%

1-Methyl-3-phenylpropylamine

1-Methyl-3-phenylpropylamine 98%
Synonym(s):
3-Amino-1-phenylbutane, 4-Phenyl-2-butylamine
Linear Formula:
C6H5CH2CH2CH(NH2)CH3
CAS Number:
Molecular Weight:
149.23
EC Number:

Products

Shipping
Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing