Skip to Content
Merck
1-Fluoro-2,4,6-trimethylpyridinium tetrafluoroborate ≥95%

1-Fluoro-2,4,6-trimethylpyridinium tetrafluoroborate

1-Fluoro-2,4,6-trimethylpyridinium tetrafluoroborate ≥95%
Synonym(s):
1-Fluoro-sym-collidinium tetrafluoroborate
Empirical Formula (Hill Notation):
C8H11BF5N
CAS Number:
Molecular Weight:
226.98

Products

Shipping
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing