Skip to Content
Merck
1-Deoxynojirimycin analytical standard

1-Deoxynojirimycin

1-Deoxynojirimycin analytical standard
Synonym(s):
1,5-Dideoxy-1,5-imino-D-sorbitol
Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17

Products

Shipping
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing