Skip to Content
Merck
1-Aza-18-crown-6 ≥98.0% (NT)

1-Aza-18-crown-6

1-Aza-18-crown-6 ≥98.0% (NT)
Synonym(s):
1,4,7,10,13-Pentaoxa-16-azacyclooctadecane
Empirical Formula (Hill Notation):
C12H25NO5
CAS Number:
Molecular Weight:
263.33
EC Number:

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing