
1-Amino-3-(aminomethyl)-3,5,5-trimethylcyclohexane

Synonym(s):
5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans, Isophorone diamine
Linear Formula:
H2NC6H7(CH3)3CH2NH2
CAS Number:
Molecular Weight:
170.30
Products
Shipping
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing