Skip to Content
Merck
17α,20β-Dihydroxy-4-pregnen-3-one crystalline

17α,20β-Dihydroxy-4-pregnen-3-one

17α,20β-Dihydroxy-4-pregnen-3-one crystalline
Synonym(s):
17α,20β-Dihydroxyprogesterone, 17α-Hydroxy-20β-dihydroprogesterone, 4-Pregnene-17α,20β-diol-3-one, Maturation Inducing hormone (MIF)
Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing