Skip to Content
Merck
1,4-Dimethoxybenzene analytical standard

1,4-Dimethoxybenzene

1,4-Dimethoxybenzene analytical standard
Synonym(s):
p-Methoxyanisole, Dimethylhydroquinone, Hydroquinone dimethyl ether, NSC 7483
Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing