Skip to Content
Merck
1,2-Propanediol tested according to Ph. Eur.

1,2-Propanediol

1,2-Propanediol tested according to Ph. Eur.
Synonym(s):
1,2-Propanediol, Propylene glycol
Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique