Skip to Content
Merck
1,2-Di(4-pyridyl)ethylene 97%

1,2-Di(4-pyridyl)ethylene

1,2-Di(4-pyridyl)ethylene 97%
Synonym(s):
1,2-Bis(4-pyridyl)ethylene, 4,4′-Vinylenedipyridine
Empirical Formula (Hill Notation):
C12H10N2
CAS Number:
Molecular Weight:
182.22
EC Number:

Products

Shipping
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing