1,1,3,3,5,5,7,7,9,9-Decamethylpentasiloxane
Synonym(s):
1,9-Dihydrodecamethylpentasiloxane
Empirical Formula (Hill Notation):
C10H32O4Si5
CAS Number:
Molecular Weight:
356.79
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing