Skip to Content
Merck
10-Bromo-1-decanol technical grade, 90%

10-Bromo-1-decanol

10-Bromo-1-decanol technical grade, 90%
Synonym(s):
Decamethylene bromohydrin
Linear Formula:
Br(CH2)10OH
CAS Number:
Molecular Weight:
237.18
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing