Skip to Content
Merck
All Photos(1)

Key Documents

TA9H98771D49

Targetmol Chemicals Inc.

ZEN-3862

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€37.28
5 MG
€87.33
10 MG
€136.32
25 MG
€240.69
50 MG
€339.74
100 MG
€455.82
200 MG
€631.55

Vendor SKU: T13394-1MG

€37.28


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
1 MG
€37.28
5 MG
€87.33
10 MG
€136.32
25 MG
€240.69
50 MG
€339.74
100 MG
€455.82
200 MG
€631.55

About This Item

CAS Number:
Molecular Weight:
340.35

Vendor SKU: T13394-1MG

€37.28


Ships in 21 days - from Aldrich Partner.

Iupac Name

5-(3,5-dimethylisoxazol-4-yl)-1-(3-fluoro-4-(oxiran-2-yl)benzyl)pyridin-2(1H)-one

Inchi Code

1S/C19H17FN2O3/c1-11-19(12(2)25-21-11)14-4-6-18(23)22(9-14)8-13-3-5-15(16(20)7-13)17-10-24-17/h3-7,9,17H,8,10H2,1-2H3

InChI key

VQHDSZCBMWYGEK-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

97.1%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/ZEN-3862

MSDS

https://www.targetmol.com/compound/ZEN-3862


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service