Skip to Content
Merck
All Photos(1)

Key Documents

T9643

Sigma-Aldrich

DL-allo-Threonine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO3
CAS Number:
Molecular Weight:
119.12
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

C[C@@H](O)[C@@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3-/m1/s1

InChI key

AYFVYJQAPQTCCC-PWNYCUMCSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Dückers et al.
Applied microbiology and biotechnology, 88(2), 409-424 (2010-08-05)
Threonine aldolases (TAs) constitute a powerful tool for catalyzing carbon-carbon bond formations in synthetic organic chemistry, thus enabling an enantio- and diastereoselective synthesis of beta-hydroxy-alpha-amino acids. Starting from the achiral precursors glycine and an aldehyde, two new stereogenic centres are
Hisashi Muramatsu et al.
Applied microbiology and biotechnology, 90(1), 159-172 (2010-12-31)
We discovered a D-phenylserine deaminase that catalyzed the pyridoxal 5'-phosphate (PLP)-dependent deamination reaction from D-threo-phenylserine to phenylpyruvate in newly isolated Arthrobacter sp. TKS1. The enzyme was partially purified, and its N-terminal amino acid sequence was analyzed. Based on the sequence
B S Bhavani et al.
The FEBS journal, 275(18), 4606-4619 (2008-08-14)
Serine hydroxymethyltransferase (SHMT) from Bacillus stearothermophilus (bsSHMT) is a pyridoxal 5'-phosphate-dependent enzyme that catalyses the conversion of L-serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. In addition, the enzyme catalyses the tetrahydrofolate-independent cleavage of 3-hydroxy amino acids and transamination. In
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service