Skip to Content
Merck
All Photos(1)

Key Documents

SML1263

Sigma-Aldrich

Arylquin 1

≥98% (HPLC)

Synonym(s):

3-(2-Fluorophenyl)-N7,N7-dimethylquinoline-2,7-diamine)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€84.30
25 MG
€277.00

€84.30


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 MG
€84.30
25 MG
€277.00

About This Item

Empirical Formula (Hill Notation):
C17H16FN3
CAS Number:
Molecular Weight:
281.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€84.30


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC2=CC(N(C)C)=CC=C2C=C1C3=CC=CC=C3F

InChI

1S/C17H16FN3/c1-21(2)12-8-7-11-9-14(17(19)20-16(11)10-12)13-5-3-4-6-15(13)18/h3-10H,1-2H3,(H2,19,20)

InChI key

RHAQINSYYSNKKI-UHFFFAOYSA-N

Biochem/physiol Actions

Arylquin 1 is a potent secretagogue of the tumor suppressor protein prostate apoptosis response-4 (Par-4), selectively inducing apoptosis in a variety of cancer cells. Arylquin 1 acts by binding to vimentin, displacing Par-4 from vimentin for secretion. Arylquin 1 produced a dose-dependent secretion of Par-4 at nanomolar concentrations.
Arylquin 1 is a potent secretagogue of the tumor suppressor protein prostate apoptosis response-4 (Par-4).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service