Skip to Content
Merck
All Photos(5)

Key Documents

SAB1402803

Sigma-Aldrich

Monoclonal Anti-HEXA antibody produced in mouse

clone 3D1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC99608, TSD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€481.00

€481.00


Estimated to ship on14 April 2025



Select a Size

Change View
100 μG
€481.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€481.00


Estimated to ship on14 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~84.3 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HEXA(3073)

General description

Hexosaminidase subunit alpha (HEXA) gene is located on the human chromosome 15q23-q24. HEXA is found to be associated with the external leaflet of plasma membrane of human fibroblasts. α and β subunits of HEXA have 57% of similarity in their amino acid sequences.

Immunogen

HEXA (AAH18927, 1 a.a. ~ 529 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MTSSRLWFSLLLAAAFAGRATALWPWPQNFQTSDQRYVLYPNNFQFQYDVSSAAQPGCSVLDEAFQRYRDLLFGSGSWPRPYLTGKRHTLEKNVLVVSVVTPGCNQLPTLESVENYTLTINDDQCLLLSETVWGALRGLETFSQLVWKSAEGTFFINKTEIEDFPRFPHRGLLLDTSRHYLPLSSILDTLDVMAYNKLNVFHWHLVDDPSFPYESFTFPELMRKGSYNPVTHIYTAQDVKEVIEYARLRGIRVLAEFDTPGHTLSWGPGIPGLLTPCYSGSEPSGTFGPVNPSLNNTYEFMSTFFLEVSSVFPDFYLHLGGDEVDFTCWKSNPEIQDFMRKKGFGEDFKQLESFYIQTLLDIVSSYGKGYVVWQEVFDNKVKIQPDTIIQVWREDIPVNYMKELELVTKAGFRALLSAPWYLNRISYGPDWKDFYVVEPLAFEGTPEQKALVIGGEACMWGEYVDNTNLVPRLWPRAGAVAERLWSNKLTSDLTFAYERLSHFRCELLRRGVQAQPLNVGFCEQEFEQT

Biochem/physiol Actions

Hexosaminidase subunit alpha (HEXA) degrades the carbohydrate moieties of glycoproteins, glycolipids and proteoglycans. Mutations in the α chain of HEXA impairs folding, trafficking and enzyme assembly. Lysosomal β-hexosaminidase α subunit is downregulated in prostate cancer cells due to hypermethylation of gene promoter.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of plasma membrane associated mature beta-hexosaminidase A, active towards GM2 ganglioside, in human fibroblasts
Mencarelli S, et al.
Febs Letters, 579(25), 5501-5506 (2005)
The X-ray crystal structure of human $\beta$-hexosaminidase B provides new insights into Sandhoff disease
Maier T, et al.
Journal of Molecular Biology, 328(3), 669-681 (2003)
Tay-Sachs disease mutations in HEXA target the alpha chain of hexosaminidase A to endoplasmic reticulum-associated degradation
Dersh D, et al.
Molecular Biology of the Cell, 27(24), 3813-3827 (2016)
Production of recombinant beta-hexosaminidase A, a potential enzyme for replacement therapy for Tay-Sachs and Sandhoff diseases, in the methylotrophic yeast Ogataea minuta
Akeboshi H, et al.
Applied and Environmental Microbiology, 73(15), 4805-4812 (2007)
Hypermethylation contributes to down-regulation of lysosomal beta-hexosaminidase alpha subunit in prostate cancer cells
Costanzi E, et al.
Biochimie, 101, 75-82 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service